Learn More
Apexbio Technology LLC Cephalexin 15686-71-2 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B16925.1
Encompass_Preferred
Cephalexin (15686-71-2) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to disrupt the bacterial cell wall synthesis thereby compromising bacterial cell wall integrity Cephalexin exerts its biological activity primarily through inhibition of peptidoglycan layer assembly by binding to PBPs In in vitro studies Cephalexin demonstrates broad-spectrum antibacterial inhibition with IC50 values typically observed in the low-micromolar concentration range depending on bacterial species and experimental conditions Based on these pharmacological properties Cephalexin holds research potential in studying bacterial infections such as pneumonia streptococcal pharyngitis and bacterial endocarditis as well as in antimicrobial susceptibility assays resistance mechanism analysis and antibiotic interaction studies
Catalog No. 50-000-96526