Learn More
Medchemexpress LLC Cephalexin monohydrate | 23325-78-2 | 99.6% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0200B5G
Encompass_Preferred
Cephalexin monohydrate is a potent, orally active, new semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum. It demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria. It targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly and is used for research related to pneumonia, strep throat, and bacterial endocarditis.
  • Potent, orally active new semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Inhibits a wide variety of gram-positive and gram-negative bacteria
  • Disrupts polymer peptidoglycan biogenesis by inactivating penicillin-binding proteins (PBPs)
  • Used for research of pneumonia, strep throat, and bacterial endocarditis
Catalog No. 50-004-07388