Learn More
Alkali Scientific Cephalexin, Monohydrate, 5 Grams
Supplier: Alkali Scientific CEP001CP
Cephalexin monohydrate is a first-generation cephalosporin antibiotic supplied in a 5 gram laboratory-grade format for research and microbiological applications. It functions by inhibiting bacterial cell wall synthesis, leading to the disruption of bacterial growth and replication. This compound is primarily effective against Gram-positive bacteria and selected Gram-negative organisms. It is commonly used in laboratory testing, antimicrobial susceptibility studies, and preparation of selective culture media. The monohydrate form ensures stability and reliable handling during experimental procedures. Cephalexin is widely used in pharmaceutical research, microbiology laboratories, and analytical applications requiring controlled antibacterial activity.