Learn More
Medchemexpress LLC Cephalexin (Standard) | 15686-71-2 | 99.99% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0200R50MG
Encompass_Preferred
Cephalexin (Standard) is the analytical standard of Cephalexin. It is a potent, orally active semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum, effective against a wide variety of gram-positive and gram-negative bacteria. It targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly and is used for research into conditions like pneumonia, strep throat, and bacterial endocarditis.
  • Intended for research and analytical applications
  • Potent, orally active semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Active against gram-positive and gram-negative bacteria
  • Targets penicillin-binding proteins (PBPs)
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-07339