Learn More
TARGETMOL CHEMICALS INC Cephalosporin C zinc salt 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T714250MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Cephalosporin C zinc salt was a potent inhibitor of SAMHD1 (IC50 1.1 ± 0.1 uM), and this inhibition was largely attributable to the presence of zinc. Purity 98%
Catalog No. 50-287-8203