Learn More
Medchemexpress LLC Cephalosporin C zinc salt | 59143-60-1 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1299A100MG
Encompass_Preferred
Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. It also demonstrates moderate anti-orthopoxvirus activity. It is 200-fold more potent than the Na+ salt form of Cephalosporin C (ICC50CC-Na = 213 ± 30 μM) in vitro, making it suitable for research use only, not for sale to patients.
  • Potent inhibitor of SAMHD1 with an IC50 of 1.1 μM
  • 200-fold more potent than the Na+ salt form in vitro
  • Moderate anti-orthopoxvirus activity
Catalog No. 50-004-12002