Learn More
Medchemexpress LLC Cephalosporin C zinc salt | 59143-60-1 | 99.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1299A5MG
Encompass_Preferred
Cephalosporin C zinc salt is a potent inhibitor of SAMHD1, with an IC50 of 1.1 μM. It exhibits moderate anti-orthopoxvirus activity and is 200-fold more potent than the Na+ salt form of Cephalosporin C.
  • Potent inhibitor of SAMHD1
  • IC50 of 1.1 μM
  • Moderate anti-orthopoxvirus activity
  • 200-fold more potent than the Na+ salt form of Cephalosporin C
Catalog No. 50-004-11598