Learn More
Medchemexpress LLC Cephalosporin C zinc salt | 59143-60-1 | 99.31% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1299A50MG
Encompass_Preferred
Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. It also exhibits moderate anti-orthopoxvirus activity. In vitro, Cephalosporin C zinc salt demonstrates an IC50 of 1.1 ± 0.1 μM for SAMHD1, making it 200-fold more potent than the Na+ salt form of Cephalosporin C.
  • Potent inhibitor of SAMHD1 (IC50 of 1.1 μM).
  • Moderate anti-orthopoxvirus activity.
Catalog No. 50-004-13614