Learn More
Medchemexpress LLC Cephalothin | 153-61-7 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB1275A200MG
Encompass_Preferred
Cephalothin (Standard) is the analytical standard of Cephalothin, intended for research and analytical applications. It is a semi-synthetic cephalosporin and beta-lactam antibiotic. It interferes with the cross-linking of peptidoglycan in the cell wall, hindering bacterial cell wall synthesis. It can be used in hematological and nephrotoxicity studies and is commonly employed in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Semi-synthetic cephalosporin and beta-lactam antibiotic
  • Inhibits class C β-lactamase AmpC
  • Interferes with bacterial cell wall synthesis
  • Exhibits antibacterial activity against a variety of bacteria
  • Suitable for hematological and nephrotoxicity studies
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-13122