Learn More
Medchemexpress LLC Cephalothin (sodium) | 58-71-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB127510MM/1ML
Encompass_Preferred
Cephalothin sodium is a semi-synthetic cephalosporin and beta-lactam antibiotic. It functions by inhibiting class C β-lactamase AmpC and interfering with the cross-linking of peptidoglycan in the bacterial cell wall, thereby hindering normal cell wall synthesis. This substance exhibits antibacterial activity against various bacteria and is suitable for use in hematological and nephrotoxicity studies.
  • Semi-synthetic cephalosporin antibiotic
  • Beta-lactam antibiotic
  • Inhibits class C β-lactamase AmpC
  • Interferes with bacterial cell wall synthesis
  • Demonstrates antibacterial activity against various bacteria
  • Suitable for hematological and nephrotoxicity studies
Catalog No. 50-004-12904