Learn More
Avanti Polar Lipids CER3(D9) 5MG
Supplier:  Avanti Polar Lipids 791923P5MG
Encompass
N-palmitoyl(d9) D-ribo-phytosphingosine, 5mg | Purity>99% | CAS Number 2260669-53-0 | Formula Weight - 564.971
Catalog No. NC2004350