Learn More
Medchemexpress LLC CER8-d9 | 2260670-29-7 | 99.0% | 562.96 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY146941S1MG
Encompass_Preferred
CER8-d9 is deuterium labeled CER8, intended for research use only. It serves as a tracer or internal standard for quantitative analysis. Deuteration, the process of incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is used for quantitation during drug development and can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Affects pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-59870