Learn More
Andwin Scientific Buret Support Stand White porcelain base supports a 3cm diameter aluminum rod. Base area: 33x17cm.
SDP
Supplier:  Andwin Scientific 17683054
Encompass_Preferred
Buret Support Stand White porcelain base supports a 3cm diameter aluminum rod. Base area: 33x17cm.
Catalog No. NC9055174