Learn More
Fine Science Tools CERAMIC BLADES
SDP
Supplier:  Fine Science Tools 1002545
Encompass
Product: Ceramic Blades - Angled/8mm Cutting Edge/ Blade Shape: Angled/ Cutting Edge: 8mm/ Thickness: 0.5mm/ Alloy / Material: Ceramic
Catalog No. NC9964554