Learn More
Thermo Scientific Reactor CHNS/NCS/S
Supplier:  Thermo Scientific 46802015
Encompass
Pre-packed combustion reactor for elemental analysis of solid samples using CHNS/NCS/S techniques to rapidly and accurately determine carbon, hydrogen, nitrogen, sulfur, and oxygen content in organic and inorganic samples by sealing oxidation reagents inside inert material, loading samples into reactor and combusting at high temperatures to convert elements into simple gases measured by thermal conductivity detector for improved precision, accuracy and lab safety.
Catalog No. 50-134-7431