Learn More
TA Instruments Ceramic Sample Pan 100 uL, 3/pk
Supplier:  TA Instruments 957329.903
Encompass
DTGA/Q5000 IR Ceramic Pans, 100 µL (pkg. of 3): These pans are available only for the Q5000 IR. Intended for operation from ambient to 1200 °C.
Catalog No. NC1258946