Learn More
Perkin Elmer US LLC Ceramic Sample Pans - Qty. 45
Supplier:  Perkin Elmer US LLC N5200045
Encompass_Preferred
These ceramic sample pans are used with the TGA 4000, STA 6000/8000 and in DSC upgrade kits. The TGA 4000 features a top loading balance for easy sample loading and unloading. The furnace and balance are isolated from operators to minimize maintenance, and its fast cooling reduces cycle times. The STA 6000 provides high performance, reliability, and productivity with an advanced innovative sensor technology. The instrument also measures both sample and reference materials simultaneously to yield greater accuracy and higher quality, and is designed for both routine and research applications.
Catalog No. 50-211-1358