Learn More
Medchemexpress LLC Ceramide 3 | 34354-88-6 | MFCD02259296 | 98.0% | 583.97 | C36H73NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1415825MG
Encompass_Preferred
Ceramide 3 (N-stearoyl phytosphingosine) is a research-grade phytoceramide used in biochemical assays to investigate skin barrier function, cell signaling, apoptosis, and cell differentiation. Supplied as a purified lipid powder for laboratory research, it is intended for research use only.
  • High purity (98.0%) suitable for analytical and biological studies.
  • Well-characterized chemical identity (CAS 34354-88-6) for traceability.
  • Compatible with common solvents and assay workflows.
  • Useful for skin barrier, signaling, and apoptosis research applications.
  • Available in small pack sizes for controlled experimental use.
  • Datasheet and safety data sheet available for handling and storage guidance.
Catalog No. 50-002-23293