Learn More
Cayman Chemical Cerdulatinib, 1198300-79-6, 10 mg
SDP
Supplier:  Cayman Chemical 2007610
Encompass_Preferred
A dual inhibitor of Syk and JAK (IC50s = 32, 12, 6, 8, and 0.5 nM for Syk, JAK1, JAK2, JAK3, and TYK2, respectively); exhibits nanomolar inhibition of other kinases; inhibits JAK1/3-mediated STAT phosphorylation with IC50 values ranging from 0.16 to 1 μM; antitumor activity with IC50 values ranging from 0.05 to 2.1 μM against DLBCL tumor cell lines
Catalog No. NC1390832