Learn More
Medchemexpress LLC Cerevisterol | 516-37-0 | 1 MG

Supplier: Medchemexpress LLC HYN35711MG
Cerevisterol is a steroid isolated from the fruiting bodies of *Agaricus blazei*. It is intended for research use only and not for sale to patients. It has demonstrated cytotoxicity against human A549, HCT-15, SK-MEL-2, SK-OV-3, and XF498 cells.
- Molecular weight: 430.66
- Formula: C28H46O3
- Appearance: Solid
- Color: White to off-white
- Purity: 99.88%
- Structure classification: Steroids
- Initial source: Microorganisms, endophytic fungi, a *Fusarium solani* strain
- Cytotoxic against human A549, HCT-15, SK-MEL-2, SK-OV-3, and XF498 cells
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.