Learn More
CELLULAR TECHNOLOGY LTD CERI-MHC CLASS I CNTRL PEPTIDE
SDP
Supplier:  CELLULAR TECHNOLOGY LTD CTLCERI300
Encompass
Peptide Pool consisting of 124 class I restricted epitopes from Cytomegalovirus (30 peptides) , Epstein Barr Virus (59 peptides), Human Respiratory Syncytial Virus (11 peptides), and Influenza A Virus (24 peptides). CELLULAR TECHNOLOGY LTD mfg# CTL-CERI-300
Catalog No. NC2031430