Learn More
CERILLIANT™ APINACA (AKB-48), 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Worldwide Glass Resource 26608
Catalog No. 01-001-987
Only null left
Add to Cart
Description
Description
APINACA, also known as AKB-48, is an aminoalkylindazole synthetic cannabinoid with a high affinity for the CB1 receptor. APINACA has been found in novel psychoactive 'smoking blends' sold through the internet. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS or LC/MS synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing applications.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation