Learn More
Selleck Chemical LLC Ceritinib dihydrochloride S4967-25mg
SDP
Supplier:  Selleck Chemical LLC S496725MG
Encompass_Preferred
Ceritinib (Zykadia LDK378) dihydrochloride is a selective orally bioavailable and ATP-competitive inhibitor of ALK with IC50 of 0 2 nM Ceritinib dihydrochloride also inhibits InsR IGF-1R and STK22D with IC50 of 7 nM 8 nM and 23 nM respectively Ceritinib exhibits antitomor activity
Catalog No. 50-311-0869