Learn More
Strem, An Ascensus Company CAS# 10060-10-3. 50g. Cerium(IV) fluoride, anhydrous, 99%. MFCD00049422
SDP
Supplier:  Strem, An Ascensus Company 58050050G
Encompass
CAS# 10060-10-3. 50g. Cerium(IV) fluoride, anhydrous, 99%. MFCD00049422. Molecular Weight: 216.13. Molecular Formula: CeF4. Color/form: white pwdr. Strem# 58-0500. http://www.strem.com/catalog/v/58-0500/
Catalog No. 50-901-12760