Learn More
Strem, An Ascensus Company CAS# 1306-38-3. 25g. Cerium(IV) oxide nanopowder. MFCD00010933
SDP
Supplier:  Strem, An Ascensus Company 58140025G
Encompass
CAS# 1306-38-3. 25g. Cerium(IV) oxide nanopowder. MFCD00010933. Molecular Weight: 172.12. Molecular Formula: CeO2. Color/form: yellow pwdr. Strem# 58-1400. http://www.strem.com/catalog/v/58-1400/
Catalog No. 50-901-12767