Learn More
Waters Corp Sep-Pak Vac Cartridge, Alumina A, 6 cc, 1 g, 30/box
Supplier:  Waters Corp 186004604
Encompass_Preferred
These disposable solid-phase extraction (SPE) devices are designed for sample preparation, offering convenience and reproducibility. They are manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines for Class 1 Medical Devices. This specific cartridge features Alumina A chemistry.
  • Convenient and reproducible
  • Disposable solid-phase extraction (SPE) device
  • Manufactured in an ISO 13485 and ISO 9002-registered facility in compliance with cGMP guidelines for Class 1 Medical Devices
  • Used for sample preparation
Catalog No. 50-791-152