Learn More
Strem, An Ascensus Company CAS# 7787-69-1. 50g. Cesium bromide (99.9%-Cs). MFCD00010954
SDP
Supplier:  Strem, An Ascensus Company 93554150G
Encompass
CAS# 7787-69-1. 50g. Cesium bromide (99.9%-Cs). MFCD00010954. Molecular Weight: 212.81. Molecular Formula: CsBr. Color/form: white xtl. Strem# 93-5541. http://www.strem.com/catalog/v/93-5541/
Catalog No. 50-901-15770