Learn More
Selleck Chemical LLC Cesium chloride S3642-25mg
SDP
Supplier:  Selleck Chemical LLC S364225MG
Encompass_Preferred
Cesium chloride (CsCl) is the inorganic compound It is widely used medicine structure in isopycnic centrifugation for separating various types of DNA and is a reagent in analytical chemistry
Catalog No. 50-310-8371