Learn More
Medchemexpress LLC Cesium chloride | 7647-17-8 | MFCD00010955 | 99.9% | 168.36 g·mol⁻¹ | CsCl | 500 G

Supplier: Medchemexpress LLC HY107754500G
Cesium chloride (CsCl) is an inorganic salt used in research as a potassium channel blocker and for biochemical and electrophysiological studies. It is supplied as a high-purity solid for laboratory use; handle according to safety data sheets and store dry at room temperature.
- CAS number 7647-17-8.
- Chemical formula CsCl.
- Molecular weight 168.36 g·mol⁻¹.
- Purity approximately 99.9%.
- Store dry at room temperature; in solution keep frozen for long-term storage.
- Suitable for electrophysiology and ion transport studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.