Learn More
Research Products International Corp Cesium Chloride, Optical Grade, 1 Kilogram
SDP
Supplier:  Research Products International Corp C680501000.0
Encompass_Preferred
Optical Density: 50% w/w solution in 1cm path cell at 260nm is <0.02.
Catalog No. 50-213-260
 Restricted Item