Learn More
Alkali Scientific Cesium Chloride, Optical Grade, 250 Grams
SDP
Supplier:  Alkali Scientific CES004CP
Encompass_Preferred
Cesium Chloride, optical grade, is a high-purity reagent supplied in a 250-gram package. It is widely used in research applications involving density gradient centrifugation and in the preparation of cesium ion solutions for various chemical and biological assays. Cesium chloride plays a critical role in the separation of nucleic acids and proteins due to its high solubility and the ability to form dense solutions. It is also used in the production of cesium vapor lamps and in certain optical devices. This 250g supply provides a convenient quantity for laboratories requiring precise, high-purity cesium chloride for experimental or analytical use.
Catalog No. 50-006-60384