Learn More
Strem, An Ascensus Company CAS# 7789-17-5. 100g. Cesium iodide (99%-Cs). MFCD00010962
SDP
Supplier:  Strem, An Ascensus Company 935524100G
Encompass
CAS# 7789-17-5. 100g. Cesium iodide (99%-Cs). MFCD00010962. Molecular Weight: 259.81. Molecular Formula: CsI. Color/form: white xtl. Strem# 93-5524. http://www.strem.com/catalog/v/93-5524/
Catalog No. 50-901-15761