Learn More
Medchemexpress LLC Cetearyl alcohol | 67762-27-0 | MFCD01076350 | 99.1% | 512.93 g/mol | C34H72O2 | 250 G
SDP
Supplier:  Medchemexpress LLC HYW422419250G
Encompass_Preferred
Cetearyl alcohol is a mixed fatty alcohol (cetyl and stearyl alcohols) used as a multifunctional excipient and cosmetic ingredient to soften, thicken, emulsify, and stabilize formulations. It is supplied as a white to off-white solid and is commonly used in topical and oral formulations for its emollient and texture-modifying properties.
  • Acts as an emollient to soften and hydrate skin
  • Functions as a thickener and viscosity modifier
  • Serves as a nonionic emulsifier in many formulation types
  • Improves formulation stability and sensory texture
  • Compatible with common cosmetic and pharmaceutical ingredients
  • Soluble in DMSO at approximately 25 mg/mL with sonication
Catalog No. 50-002-81337