Learn More
Medchemexpress LLC Cetearyl alcohol | 67762-27-0 | MFCD04113584 | 99.1% | 512.9 g/mol | C34H72O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW42241950G
Encompass_Preferred
Cetearyl alcohol is a pharmaceutical excipient, a blended fatty alcohol (cetyl and stearyl alcohols) supplied as a white, waxy solid. It functions as an emollient, emulsifier, and thickening agent to improve texture, stability, and processing of topical formulations and other dosage forms. Certificate of analysis (COA), safety data sheet (SDS), and handling information are available.
  • High purity (99.06%) suitable for formulation use.
  • Acts as an emollient, emulsifier, and thickener.
  • Improves stability, texture, and processability of formulations.
  • Available in multiple package sizes including 25 g, 50 g, and larger.
  • Certificate of analysis and safety data sheet available for quality and compliance.
  • White waxy solid, easy to weigh and incorporate into formulations.
Catalog No. 50-002-80738