Learn More
Medchemexpress LLC Cetearyl alcohol | 67762-27-0 | MFCD04113584 | 99.1% | 512.93 g/mol | C34H72O2 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW42241925G
Encompass_Preferred
Cetearyl alcohol is a white to off-white solid fatty alcohol mixture used as a pharmaceutical excipient and cosmetic emulsifier. It acts as an emollient, thickener, and co-emulsifier for formulation and research applications, and is supplied at high purity for laboratory use.
  • High purity (99.06%).
  • Mixture of C16-C18 fatty alcohols (cetyl and stearyl alcohol).
  • White to off-white solid with a waxy texture.
  • Functions as emollient, thickener, and co-emulsifier in formulations.
  • Supplied in laboratory-friendly package sizes for formulation work.
Catalog No. 50-002-81330