Learn More
Medchemexpress LLC Cetirizine-d4 dihydrochloride | 83881-52-1 | 465.83 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17042S5MG
Encompass_Preferred
Cetirizine-d4 dihydrochloride is a deuterium-labeled variant of Cetirizine, which is a second-generation antihistamine. It functions as a specific, orally active, and long-acting antagonist of the histamine H1-receptor. This compound exhibits antiallergic properties and can inhibit eosinophil chemotaxis during allergic responses. It is intended for research purposes only, particularly as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-85732