Learn More
Medchemexpress LLC Cetirizine-d4 dihydrochloride | 83881-52-1 | ≥98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17042S10MG
Encompass_Preferred
Cetirizine-d4 dihydrochloride is a deuterium-labeled research standard of cetirizine, supplied for analytical and laboratory use. The compound is provided as the dihydrochloride salt and is intended for use as an internal standard or tracer in mass spectrometry and chromatographic assays.
  • Deuterium-labeled for mass spectrometry and tracer studies.
  • Dihydrochloride salt for consistent handling and stability.
  • High chemical purity (≥98.0%).
  • Supplied in small milligram quantities suitable for analytical standards.
  • Suitable as an internal standard for HPLC-MS and quantitative assays.
Catalog No. 50-002-95315