Learn More
Medchemexpress LLC Cetirizine-d8 | 774596-22-4 | 96.5% | 396.94 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17042S15MG
Encompass_Preferred
Cetirizine-d8 is a deuterium labeled Cetirizine. Cetirizine is a second-generation antihistamine and the carboxylated metabolite of hydroxyzine. It is a specific, orally active, and long-acting histamine H1-receptor antagonist. Cetirizine exhibits antiallergic properties and inhibits eosinophil chemotaxis during the allergic response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compounds
  • Internal standard for clinical mass spectrometry
  • Isotope-labeled inhibitors
  • Isotope-labeled metabolites
  • Deuterium (2H, D) stable isotope-labeled inhibitors
  • Stable isotope-labeled inhibitors for use in in vivo cell experiments
  • Therapeutic drug monitoring: TDM
Catalog No. 50-003-86096