Learn More
Cayman Chemical CetIrIzInhydrochlorIde 10g
SDP
Supplier:  Cayman Chemical 1968610 G
Encompass_Preferred
A selective histamine H1 receptor antagonist (Ki = 10 nM); inhibits eosinophil chemotaxis and LTB4 release independent from H1 antagonism; inhibits aerosol histamine-induced bronchospasm in guinea pigs (ED50 = 100 μg/kg, p.o.)
Catalog No. 50-298-4109