Learn More
Sigma Aldrich Fine Chemicals Biosciences CETP Activity Assay Kit Su
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK1061KT
Encompass
Cholesteryl ester transfer protein (CETP) is present in normal human plasma. The protein transfers neutral lipids from high density lipoproteins (HDL) to very low density lipoprotein (VLDL) and low density lipoprotein (LDL). CETP plays an important role in lipoprotein metabolism and influences the reverse cholesterol transport pathway. The method is useful for measuring CETP activity in plasma or serum in all species that express CETP.
Catalog No. 50-290-7624