Learn More
Medchemexpress LLC Cetuximab PBS 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9S00081MG
Encompass_Preferred
Cetuximab (PBS) is a human IgG1 monoclonal antibody that can inhibit EGFR The SPR method measured the Kd value of Cetuximab for EGFR to be 0 201 nM Cetuximab has a highly effective anti-tumor effect[1]
Catalog No. 50-003-21977