Learn More
Apexbio Technology LLC Cetylpyridinium Chloride 123-03-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16945.1
Encompass_Preferred
Cetylpyridinium chloride (CAS 123-03-5) is a small-molecule inhibitor targeting microbial cell membranes It is designed to disrupt bacterial lipid bilayers thereby altering membrane permeability and inhibiting microbial viability Cetylpyridinium chloride exerts its biological activity primarily through membrane destabilization via positively charged cetylpyridinium ions interacting with negatively charged microbial surfaces In in vitro studies cetylpyridinium chloride demonstrates inhibitory activity with IC50 values ranging approximately between 0 5 to 10 M depending on microbial strain type and experimental conditions Based on these pharmacological properties cetylpyridinium chloride holds research potential in evaluating oral microbial inhibition biofilm formation interference and cell membrane-targeted antibacterial strategies
Catalog No. 50-000-94409