Learn More
Medchemexpress LLC Cevidoplenib dimesylate | 2043659-93-2 | 99.0% | 665.74 g·mol-1 | C27H35N7O9S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY109082A5MG
Encompass_Preferred
Cevidoplenib dimesylate is an orally active small-molecule inhibitor of spleen tyrosine kinase (Syk) supplied for research use. It is intended for in vitro and in vivo studies of Syk-mediated signaling, inflammation, and immune modulation, and is available in solid and solution formats to accommodate different experimental workflows.
  • High purity suitable for preclinical research.
  • Orally bioavailable Syk inhibitor with activity in cellular assays.
  • Available as solid vials in multiple sizes and as a DMSO solution.
  • Supplied as the dimesylate salt for consistent formulation and handling.
  • Compatible with common laboratory solvents, including DMSO, for stock preparation.
  • Applicable to pharmacology, immunology, and disease model studies.
Catalog No. 50-002-64477