Learn More
Cayman Chemical CevImelInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1780210 MG
Encompass_Preferred
A muscarinic receptor agonist (EC50s = 23, 48, and 63 nM for M1, M3, and M5, respectively, and >1 µM for M2 and M4); stimulates secretion by salivary glands and is useful in ameliorating xerostomia (dry mouth)
Catalog No. 50-296-6916