Learn More
Biotium CF488A Donkey Anti-Chicken IgY (IgG) (H+L), Highly Cross-Adsorbed, 2 mg/mL
Supplier:  Biotium 20166
Encompass_Preferred
Fluorescently labeled Donkey Anti-Chicken IgY (IgG) (H+L), Highly Cross-Adsorbed is prepared by labeling high quality antibody with a selection of fluorescent CF dyes and other labels. CF dyes are next-generation fluorescent dyes with advantages brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A has excitation/emission at 490/515 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.
Catalog No. 50-196-4251