Learn More
Biotium Secondary Antibody Mouse IgG Donkey CF514 0 1/EA
Supplier:  Biotium 204831MG
Encompass_Preferred
Highly cross-adsorbed donkey anti-mouse IgG (H plus L) labeled with green fluorescent CF and reg 514 dye (Ex/Em 516/548 nm) The antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human rabbit and sheep serum 1 mg lyophilized
Catalog No. 50-196-4361