Learn More
TARGETMOL CHEMICALS INC CF53 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T107735MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CF53 is a highly potent selective and orally active inhibitor of BET protein with a Ki of <1 nM Kd of 2.2 nM and an IC50 of 2 nM for BRD4 BD1. CF53 binds to both the BD1 and BD2 domains of BRD2 BRD3 BRD4 and BRDT BET proteins with high affinities very selective over non-BET bromodomain-containing proteins. CF53 shows potent anti-tumor activity both in vitro and in vivo. purity: 100%
Catalog No. 50-258-1670