Learn More
Biotium CF568 Donkey Anti-Goat IgG (H+L), Highly Cross-Adsorbed, 2 mg/mL
Supplier:  Biotium 20106
Encompass_Preferred
Fluorescently labeled Donkey Anti-Goat IgG (H+L), Highly Cross-adsorbed is prepared by labeling high quality antibody with a selection of fluorescent CF dyes and other labels. CF dyes are next-generation fluorescent dyes with advantages brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 has excitation/emission at 562/583 nm. Supplied at 2 mg/mL in PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide.
Catalog No. 50-196-4212