Learn More
Biotium CF568 Donkey Anti-Rabbit IgG (H+L), Highly Cross-Adsorbed, single label for STORM, 1 mg/mL
Supplier:  Biotium 20803500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF568 dye. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 dye has excitation/emission at 562/583 nm. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum. Single label antibodies have an average of one dye molecule per antibody molecule, which is reported to be optimal for super-resolution imaging by STORM.
Catalog No. 50-196-4371