Learn More
Biotium CF568 Tyramide
Supplier:  Biotium 92173
Encompass_Preferred
CF dye tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 dye has excitation/emission at 562/583 nm.Also see Biotium's CF Dye Tyramide Amplification Kits.
Catalog No. 50-196-4912